2,2-Bis(trifluoromethyl)-6-iodo-1,1,1-trifluorohexane ≥97%

Supplier: Matrix Scientific

012833-1G
101803-480EA 144.62 USD
101803-480
2,2-Bis(trifluoromethyl)-6-iodo-1,1,1-trifluorohexane ≥97%
2,2-Bis(trifluoromethyl)-6-iodo-1,1,1-trifluorohexane

Hazard: Irritant

Formula: C₈H₈F₉I
MDL Number: MFCD06246014
CAS Number: 883516-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR