3-Phenyl-5-isoxazolecarboxylic acid

Supplier: Matrix Scientific

Synonyms: 3-Phenylisoxazole-5-carboxylic acid

012792-500MG
101803-398EA 275.38 USD
101803-398
3-Phenyl-5-isoxazolecarboxylic acid
3-Phenyl-5-isoxazolecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₃
MW: 189.17
Melting Pt: 177…178 °C
MDL Number: MFCD01936010
CAS Number: 14442-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR