3-Imidazol-1-ylpropionic acid

Supplier: Matrix Scientific

012750-1G
101803-320EA 346.15 USD
101803-320
3-Imidazol-1-ylpropionic acid
3-Imidazol-1-ylpropionic acid

Hazard: Irritant

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Melting Pt: 157…158 °C
MDL Number: MFCD02731119
CAS Number: 18999-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR