3-(Morpholinomethyl)aniline

Supplier: Matrix Scientific

012739-500MG
101803-298EA 58.46 USD
101803-298
3-(Morpholinomethyl)aniline
3-(Morpholinomethyl)aniline

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MW: 192.26 g/mol
Boiling Pt: 184…189 °C (9 Torr)
MDL Number: MFCD03074538
CAS Number: 123207-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR