5-Thiophen-2-yl-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

012736-500MG
101803-292EA 153.85 USD
101803-292
5-Thiophen-2-yl-1H-pyrazole-3-carboxylic acid
5-Thiophen-2-yl-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂S
MW: 194.2104 g/mol
MDL Number: MFCD03030176
CAS Number: 182415-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR