3-Hydroxy-1-adamantaneacetic acid

Supplier: Matrix Scientific

Synonyms: 1-(Carboxymethyl)-3-hydroxyadamantane2-(3-Hydroxyadamantan-1-yl)acetic acid

012727-500MG
101803-274EA 23.08 USD
101803-274
3-Hydroxy-1-adamantaneacetic acid
3-Hydroxy-1-adamantaneacetic acid

Hazard: Irritant

Formula: C₁₂H₁₈O₃
MW: 210.27 g/mol
Melting Pt: 126…128 °C
MDL Number: MFCD00167797
CAS Number: 17768-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR