3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid
Supplier: Matrix Scientific
Synonyms:
3-(4-Methoxyphenyl)isoxazole-5-carboxylic acid
012722-500MG
101803-264EA
275.38
USD
101803-264
3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid
3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid
Hazard: Irritant
Formula:
C₁₁H₉NO₄ MW: 219.1900 g/mol Melting Pt: 138 °C |
MDL Number:
MFCD04124288 CAS Number: 618383-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...