2-Methylquinoline-4-carboxylic acid

Supplier: Matrix Scientific

012696-500MG
101803-226EA 24.62 USD
101803-226
2-Methylquinoline-4-carboxylic acid
2-Methylquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₂
Melting Pt: 244 °C
MDL Number: MFCD00093919
CAS Number: 634-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR