5-Nitroindole-2-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Nitro-1H-indole-2-carboxylic acid

012603-1G
101802-958EA 44.62 USD
101802-958
5-Nitroindole-2-carboxylic acid ≥98%
5-Nitroindole-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆N₂O₄
MW: 206.157 g/mol
Melting Pt: 326…328 °C
MDL Number: MFCD00236372
CAS Number: 16730-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR