1-N-Boc-4-(2-furfurylmethylamino)piperidine ≥98%

Supplier: Matrix Scientific

Synonyms: 4-[(Fur-2-ylmethyl)amino]piperidine, N1-BOC protected

012598-1G
101802-948EA 213.85 USD
101802-948
1-N-Boc-4-(2-furfurylmethylamino)piperidine ≥98%
1-N-Boc-4-(2-furfurylmethylamino)piperidine

Hazard: Irritant

Formula: C₁₅H₂₄N₂O₃
MW: 280.37 g/mol
MDL Number: MFCD06245534
CAS Number: 883516-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR