(2-Aminoethyl)benzyl carbamic acid-tert-butylester ≥98%

Supplier: Matrix Scientific

Synonyms: N-Benzylethane-1,2-diamine, N-BOC protected

012556-1G
101802-850EA 106.15 USD
101802-850
(2-Aminoethyl)benzyl carbamic acid-tert-butylester ≥98%
(2-Aminoethyl)benzyl carbamic acid-tert-butylester

Hazard: Irritant

Formula: C₁₄H₂₂N₂O₂
MW: 250.34 g/mol
MDL Number: MFCD06245501
CAS Number: 152193-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR