N-Adamantan-1-yl-2-chloroacetamide

Supplier: Matrix Scientific

012492-500MG
101802-680EA 273.85 USD
101802-680
N-Adamantan-1-yl-2-chloroacetamide
N-Adamantan-1-yl-2-chloroacetamide

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MW: 227.74 g/mol
Melting Pt: 113…115 °C
MDL Number: MFCD01733430
CAS Number: 5689-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR