Benzylmethylpiperidin-4-ylamine dihydrochloride

Supplier: Matrix Scientific

012489-500MG
101802-674EA 273.85 USD
101802-674
Benzylmethylpiperidin-4-ylamine dihydrochloride
Benzylmethylpiperidin-4-ylamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁ClN₂
MW: 240.78 g/mol
MDL Number: MFCD08445692
CAS Number: 871112-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR