4-Piperidin-1-ylmethylbenzoic acid

Supplier: Matrix Scientific

Synonyms: 4-(1-Piperidinylmethyl)-benzoic acid hydrochloride

012479-500MG
101802-654EA 273.85 USD
101802-654
4-Piperidin-1-ylmethylbenzoic acid
4-Piperidin-1-ylmethylbenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₂
MW: 219.29 g/mol
MDL Number: MFCD05240080
CAS Number: 159691-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR