4-(2-Aminothiazol-4-yl)phenol

Supplier: Matrix Scientific

Synonyms: 4-(2-Amino-4-thiazolyl)phenol

012471-500MG
101802-638EA 273.85 USD
101802-638
4-(2-Aminothiazol-4-yl)phenol
4-(2-Aminothiazol-4-yl)phenol

Hazard: Irritant

Formula: C₉H₈N₂OS
MW: 192.24 g/mol
Melting Pt: 198…200 °C
MDL Number: MFCD01624171
CAS Number: 57634-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR