2-Amino-4-methylpyrimidine-5-carboxylic acid

Supplier: Matrix Scientific

012405-500MG
101802-510EA 143.08 USD
101802-510
2-Amino-4-methylpyrimidine-5-carboxylic acid
2-Amino-4-methylpyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₆H₇N₃O₂
Melting Pt: 313…316 °C
MDL Number: MFCD05237187
CAS Number: 769-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR