(3-Nitropyrazol-1-yl)acetic acid

Supplier: Matrix Scientific

012392-500MG
101802-486EA 273.85 USD
101802-486
(3-Nitropyrazol-1-yl)acetic acid
(3-Nitropyrazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₅H₅N₃O₄
MDL Number: MFCD01622829
CAS Number: 887408-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR