Bis(3-pyridylmethyl)amine

Supplier: Matrix Scientific

Synonyms: 3,3'-Dipicolylaminealpha,alpha'-Iminodi(3-picoline)

012348-500MG
101802-402EA 273.85 USD
101802-402
Bis(3-pyridylmethyl)amine
Bis(3-pyridylmethyl)amine

CAS: 1656-94-6
MDL Number: MFCD00038043
Molecular Formula: C12H13N3
Molecular Weight: 199,26
Boiling Point: 168°C/0.55 Torr
Hazard: Irritant

Formula: C₁₂H₁₃N₃
MW: 199.26 g/mol
Boiling Pt: 168 °C (0.55 Torr)
MDL Number: MFCD00038043
CAS Number: 1656-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR