2-{[(4-Fluorophenyl)sulfonyl]amino}propanoic acid

Supplier: Matrix Scientific

012280-500MG
101802-268EA 273.85 USD
101802-268
2-{[(4-Fluorophenyl)sulfonyl]amino}propanoic acid
2-{[(4-Fluorophenyl)sulfonyl]amino}propanoic acid

Hazard: Irritant

Formula: C₉H₁₀FNO₄S
MDL Number: MFCD00664348
CAS Number: 250714-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR