2-(4-Methyl-1-piperazinyl)aniline
Supplier: Matrix Scientific
Synonyms:
1-(2-Aminophenyl)-4-methylpiperazine
012155-500MG
101802-020EA
15.38
USD
101802-020
2-(4-Methyl-1-piperazinyl)aniline
2-(4-Methyl-1-piperazinyl)aniline
Hazard: Irritant
Formula:
C₁₁H₁₇N₃ MW: 191.28 g/mol Melting Pt: 88…89 °C |
MDL Number:
MFCD04035359 CAS Number: 180605-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...