1-Methylamino-3-phenoxypropan-2-ol

Supplier: Matrix Scientific

008464-500MG
101794-728EA 258.46 USD
101794-728
1-Methylamino-3-phenoxypropan-2-ol
1-Methylamino-3-phenoxypropan-2-ol

Hazard: Irritant

Formula: C₁₀H₁₅NO₂
Melting Pt: 68…69 °C
MDL Number: MFCD01442095
CAS Number: 39631-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR