4-Oxo-4-(3-oxo-3,4-dihydro-2H-quinoxalin-1-yl)butyric acid

Supplier: Matrix Scientific

008388-500MG
101794-582EA 258.46 USD
101794-582
4-Oxo-4-(3-oxo-3,4-dihydro-2H-quinoxalin-1-yl)butyric acid
4-Oxo-4-(3-oxo-3,4-dihydro-2H-quinoxalin-1-yl)butyric acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₄
MW: 248.24 g/mol
MDL Number: MFCD02737889
CAS Number: 462068-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR