4-Amino-5-trifluoromethyl-4H-[1,2,4]triazole-3-thiol

Supplier: Matrix Scientific

008327-500MG
101794-464EA 258.46 USD
101794-464
4-Amino-5-trifluoromethyl-4H-[1,2,4]triazole-3-thiol
4-Amino-5-trifluoromethyl-4H-[1,2,4]triazole-3-thiol

Hazard: Irritant

Formula: C₃H₃F₃N₄S
MDL Number: MFCD00101076
CAS Number: 24848-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR