4-(Piperidin-1-yl)benzaldehyde

Supplier: Matrix Scientific

008324-500MG
101794-458EA 258.46 USD
101794-458
4-(Piperidin-1-yl)benzaldehyde
4-(Piperidin-1-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₅NO
MW: 189.2500 g/mol
Melting Pt: 65…66 °C
MDL Number: MFCD00778395
CAS Number: 10338-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR