1-(4-Formylphenyl)pyrrolidine
Synonyms:
4-(1-Pyrrolidino)benzaldehyde
, 4-(1-Pyrrolidinyl)benzaldehyde
008273-500MG
101794-362EA
181.9
USD
101794-362
1-(4-Formylphenyl)pyrrolidine
1-(4-Formylphenyl)pyrrolidine
Hazard: Irritant
Formula:
C₁₁H₁₃NO MW: 175.23 g/mol Melting Pt: 84…85 °C |
MDL Number:
MFCD00015469 CAS Number: 51980-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...