3,5-Dimethyl-1H-pyrazol-4-ylamine

Supplier: Matrix Scientific

Synonyms: 3,5-Dimethyl-1H-pyrazol-4-amine

008264-500MG
101794-344EA 258.46 USD
101794-344
3,5-Dimethyl-1H-pyrazol-4-ylamine
3,5-Dimethyl-1H-pyrazol-4-ylamine

Hazard: Irritant

Formula: C₅H₉N₃
MW: 111.15 g/mol
Melting Pt: 204…205 °C
MDL Number: MFCD00187666
CAS Number: 5272-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR