(2-Benzothiazolylthio)acetic acid
Supplier: Matrix Scientific
Synonyms:
2-(Benzo[d]thiazol-2-ylthio)acetic acid
, [(1,3-Benzothiazol-2-yl)sulphanyl]acetic acid
008232-500MG
101794-286EA
258.46
USD
101794-286
(2-Benzothiazolylthio)acetic acid
(2-Benzothiazolylthio)acetic acid
Hazard: Irritant
Formula:
C₉H₇NO₂S₂ MW: 225.28 g/mol Melting Pt: 156…157 °C |
MDL Number:
MFCD00227391 CAS Number: 6295-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...