3-Benzotriazol-1-ylbutyric acid

Supplier: Matrix Scientific

008231-500MG
101794-284EA 258.46 USD
101794-284
3-Benzotriazol-1-ylbutyric acid
3-Benzotriazol-1-ylbutyric acid

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₂
Melting Pt: 150 °C
MDL Number: MFCD02232744
CAS Number: 654-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR