1-Methanesulfonylpiperazine hydrochloride

Synonyms: 1-(Methylsulfonyl)piperazine hydrochloride1-(Methylsulphonyl)piperazine hydrochloride

008211-500MG
101794-244EA 181.9 USD
101794-244
1-Methanesulfonylpiperazine hydrochloride
1-Methanesulfonylpiperazine hydrochloride

Hazard: Irritant

Formula: C₅H₁₃ClN₂O₂S
MW: 200.69 g/mol
Melting Pt: 185…187 °C
MDL Number: MFCD02018964
CAS Number: 161357-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR