3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid
Supplier: Matrix Scientific
Synonyms:
3-Amino-3-(3,4-dimethoxyphenyl)propionic acid
008168-500MG
101794-154EA
258.46
USD
101794-154
3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid
3-Amino-3-(3,4-dimethoxyphenyl)propanoic acid
Hazard: Irritant
Formula:
C₁₁H₁₅NO₄ MW: 225.244 g/mol Melting Pt: 248…249 °C |
MDL Number:
MFCD00735172 CAS Number: 34841-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...