1-(2,5-Dimethoxybenzyl)piperazine hydrochloride

Supplier: Matrix Scientific

008147-500MG
101794-108EA 115.38 USD
101794-108
1-(2,5-Dimethoxybenzyl)piperazine hydrochloride
1-(2,5-Dimethoxybenzyl)piperazine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁ClN₂O₂
MDL Number: MFCD03093088
CAS Number: 374897-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR