1-(2-Methylbenzyl)piperazine hydrochloride

Supplier: Matrix Scientific

008108-500MG
101794-032EA 258.46 USD
101794-032
1-(2-Methylbenzyl)piperazine hydrochloride
1-(2-Methylbenzyl)piperazine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂
MDL Number: MFCD05656270
CAS Number: 435345-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR