1-(1H-Benzoimidazol-2-yl)ethylamine dihydrochloride

Supplier: Matrix Scientific

008100-500MG
101794-018EA 258.46 USD
101794-018
1-(1H-Benzoimidazol-2-yl)ethylamine dihydrochloride
1-(1H-Benzoimidazol-2-yl)ethylamine dihydrochloride

Hazard: Irritant

Formula: C₉H₁₃Cl₂N₃
Melting Pt: 133…138 °C
MDL Number: MFCD06799786
CAS Number: 74461-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR