(1H-Benzoimidazol-2-ylsulfanyl)acetic acid

Supplier: Matrix Scientific

008029-500MG
101793-874EA 258.46 USD
101793-874
(1H-Benzoimidazol-2-ylsulfanyl)acetic acid
(1H-Benzoimidazol-2-ylsulfanyl)acetic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂S
Melting Pt: 190 °C
MDL Number: MFCD00022673
CAS Number: 3042-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR