1-(6-Chloropyridazin-3-yl)azepane

Supplier: Matrix Scientific

008018-500MG
101793-850EA 218.46 USD
101793-850
1-(6-Chloropyridazin-3-yl)azepane
1-(6-Chloropyridazin-3-yl)azepane

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MDL Number: MFCD01460065
CAS Number: 312318-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR