[(Furan-2-carbonyl)methylamino]acetic acid

Supplier: Matrix Scientific

007945-500MG
101793-708EA 300 USD
101793-708
[(Furan-2-carbonyl)methylamino]acetic acid
[(Furan-2-carbonyl)methylamino]acetic acid

Hazard: Irritant

Formula: C₈H₉NO₄
MDL Number: MFCD00573161
CAS Number: 625408-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR