3-Methylsulfanyl[1,2,4]thiadiazol-5-ylamine

Supplier: Matrix Scientific

Synonyms: 3-(Methylthio)-1,2,4-thiadiazol-5-amine

007902-500MG
101793-624EA 258.46 USD
101793-624
3-Methylsulfanyl[1,2,4]thiadiazol-5-ylamine
3-Methylsulfanyl[1,2,4]thiadiazol-5-ylamine

Hazard: Irritant

Formula: C₃H₅N₃S₂
MW: 147.22 g/mol
Melting Pt: 142 °C
MDL Number: MFCD00100135
CAS Number: 6913-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR