1-Amino-3-(4-methoxyphenoxy)propan-2-ol

Supplier: Matrix Scientific

Synonyms: 4-(3-Amino-2-hydroxypropoxy)anisole

007892-500MG
101793-604EA 115.38 USD
101793-604
1-Amino-3-(4-methoxyphenoxy)propan-2-ol
1-Amino-3-(4-methoxyphenoxy)propan-2-ol

Hazard: Irritant

Formula: C₁₀H₁₅NO₃
MW: 197.24 g/mol
MDL Number: MFCD00205693
CAS Number: 5002-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR