2-Aminothiazole-4-carboxylic acid, ethyl ester hydrochloride
Supplier: Matrix Scientific
007879-500MG
101793-580EA
258.46
USD
101793-580
2-Aminothiazole-4-carboxylic acid, ethyl ester hydrochloride
2-Aminothiazole-4-carboxylic acid, ethyl ester hydrochloride
Hazard: Irritant
Formula:
C₆H₉ClN₂O₂S |
MDL Number:
MFCD00859544 CAS Number: 435342-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...