3-(2-Hydroxyphenylcarbamoyl)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

Supplier: Matrix Scientific

007842-500MG
101793-504EA 230.77 USD
101793-504
3-(2-Hydroxyphenylcarbamoyl)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
3-(2-Hydroxyphenylcarbamoyl)bicyclo[2.2.1]hept-5-ene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₅NO₄
MDL Number: MFCD06799404
CAS Number: 329257-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR