(1,1-Dioxotetrahydro-1-λ*6*-thiophen-3-ylamino)acetic acid

Supplier: Matrix Scientific

007729-500MG
101793-282EA 258.46 USD
101793-282
(1,1-Dioxotetrahydro-1-λ*6*-thiophen-3-ylamino)acetic acid
(1,1-Dioxotetrahydro-1-lambda*6*-thiophen-3-ylamino)acetic acid

Hazard: Irritant

Formula: C₆H₁₁NO₄S
MW: 193.22 g/mol
MDL Number: MFCD00462458
CAS Number: 51070-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR