3-Pyrid-3-ylbenzoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Pyridin-3-yl-benzoic acid

007709-500MG
101793-264EA 34.5 USD
101793-264
3-Pyrid-3-ylbenzoic acid ≥97%
3-Pyrid-3-ylbenzoic acid

Hazard: Irritant/Refrigerate

Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
MDL Number: MFCD03426512
CAS Number: 4385-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR