4-(Hydroxymethyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Hydroxymethyl)benzeneboronic acid

007693-5G
101793-246EA 46.15 USD
101793-246
4-(Hydroxymethyl)phenylboronic acid ≥95%
4-(Hydroxymethyl)phenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 73…75 °C
MDL Number: MFCD00792672
CAS Number: 59016-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR