5-Fluoro-2-methylphenylboronic acid ≥96%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-2-methylbenzeneboronic acid2-Methyl-5-fluorophenylboronic acid

007689-1G
101793-240EA 31.05 USD
101793-240
5-Fluoro-2-methylphenylboronic acid ≥96%
5-Fluoro-2-methylphenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Melting Pt: 148…149 °C
MDL Number: MFCD03095047
CAS Number: 163517-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR