(Di-n-butylamino)acetonitrile ≥97%

Supplier: Matrix Scientific

007551-100G 007551-25G
101793-028EA 73.85 USD
101793-028 101793-030
(Di-n-butylamino)acetonitrile ≥97%
(Di-n-butylamino)acetonitrile

Hazard: Irritant

Formula: C₁₀H₂₀N₂
Boiling Pt: 85 °C
Density: 0.8896
MDL Number: MFCD00019825
CAS Number: 18071-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR