3-(Methoxymethyl)phenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Methoxymethylphenylboronic acid(3-(Methoxymethyl)phenyl)boronic acid

007416-500MG
101792-700EA 76.92 USD
101792-700
3-(Methoxymethyl)phenylboronic acid ≥98%
3-(Methoxymethyl)phenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₈H₁₁BO₃
MW: 165.99 g/mol
MDL Number: MFCD03412069
CAS Number: 142273-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR