3-(Methoxycarbonyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Methoxycarbonyl)benzeneboronic acid

007413-5G
101792-694EA 13.8 USD
101792-694
3-(Methoxycarbonyl)phenylboronic acid ≥97%
3-(Methoxycarbonyl)phenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₈H₉BO₄
MW: 179.97 g/mol
Melting Pt: 136 °C
MDL Number: MFCD02093046
CAS Number: 99769-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR