4-(Benzyloxy)phenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: (4-(Benzyloxy)phenyl)boronic acid4-Benzyloxybenzeneboronic acid

007400-25G
101792-654EA 70.15 USD
101792-654
4-(Benzyloxy)phenylboronic acid ≥98%
4-(Benzyloxy)phenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₁₃H₁₃BO₃
MW: 228.06 g/mol
Melting Pt: 187…192 °C
MDL Number: MFCD01075705
CAS Number: 146631-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR