3-(Benzyloxy)phenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: (3-(Benzyloxy)phenyl)boronic acid3-Benzyloxybenzeneboronic acid

007399-5G 007399-1G
101792-646EA 11.5 USD
101792-646 101792-648
3-(Benzyloxy)phenylboronic acid ≥98%
3-(Benzyloxy)phenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₁₃H₁₃BO₃
MW: 228.06 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD02093063
CAS Number: 156682-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR