Pyridine-3-boronic acid neopentylglycol ester ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine

007389-1G
101792-632EA 21.54 USD
101792-632
Pyridine-3-boronic acid neopentylglycol ester ≥97%
Pyridine-3-boronic acid neopentylglycol ester

Hazard: Irritant/Refrigerate

Formula: C₁₀H₁₄BNO₂
MW: 191.04 g/mol
MDL Number: MFCD03425979
CAS Number: 845885-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR